Learn More
Sino Biological Recombinant Human RIOK1 Protein (Full length, GST Tag), Active 10 µg
Supplier:  Sino Biological R0410G10
Encompass_Preferred
Recombinant full-length human RIOK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-006-56715