Learn More
Sino Biological Recombinant Human SLC30A1 Protein (Nanodisc, His Tag) 20 µg
Supplier:  Sino Biological 19600H08HNA20
Encompass_Preferred
A DNA sequence encoding the Human SLC30A1(Met1-Leu507) was expressed with a polyhistidine tag at the C-terminus. This protein was prepared using our Nanodisc platform. Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.
Catalog No. 50-006-56765