Learn More
Sino Biological Recombinant Human DcR3/TNFRSF6B Protein (ECD, hFc & AVI Tag), Avi-tag Biotinylated 20 µg
Supplier:  Sino Biological 10224H41B1B20
Encompass_Preferred
A DNA sequence encoding the Human TNFRSF6B (O95407)(Met1-His300) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-006-99020