Learn More
Sino Biological Protein Human TNFRSF9
Supplier:  Sino Biological 10041H86HSG25
Encompass_Preferred
A DNA sequence encoding the human 4-1BB/CD137 (NP-001552 2) (Met 1-Gln 186) was expressed with a C-terminal polyhistidine tag The protein is site-specifically conjugated with AF 647 (Excitation Max 655 nm Emission Max 680 nm)
Catalog No. 50-001-31067