Learn More
Sino Biological Recombinant Human USP15 Protein (GST Tag), Active 500 µg
Supplier: Sino Biological U515381G500
Recombinant human USP15 (2-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.