Learn More
GenDEPOT Xpert Protease Inhibitor Cocktail Solution(100X)
Supplier:  GenDEPOT P3100020
Encompass_Preferred
The Xpert Protease Inhibitor Cocktail Solution(100X) is a specially formulated mixture of 5 protease inhibitors with broad specificity for the inhibition of various protease and esterase.
Catalog No. 50-195-9717