Learn More
Medchemexpress LLC Protein Kinase C (19-31) | 121545-65-1 | 99.7% | 1543.82 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP17465MG
Encompass_Preferred
Protein Kinase C (19-31) is a peptide inhibitor of protein kinase C (PKC). It is derived from the pseudo-substrate regulatory domain of PKCa (residues 19-31) and is utilized as a protein kinase C substrate peptide for testing PKC activity.
  • Peptide inhibitor of protein kinase C
  • Derived from PKCa (residues 19-31)
  • Used as a substrate peptide for testing protein kinase C activity
  • Purity of 99.7%
  • For research use only
Catalog No. 50-004-23793