Learn More
CPC Scientific H-Gly-Pro-Lys-Thr-Pro-Glu-Glu-Lys-Thr-Ala-Asn-Thr-Ile-Ser-Lys-Phe-Asp-Cys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PKCP035A
Encompass
ONE-LETTER SEQUENCE: GPKTPEEKTANTISKFDCMOLECULAR FORMULA: C84H136N22O30S1MOLECULAR WEIGHT:1966.2STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and Phosphatases
Catalog No. 50-211-0186