Learn More
LUNA NANOTECH INC Protein L Magnetic Beads, 1 ML
Supplier:  LUNA NANOTECH INC MGBPRL1
Encompass_Preferred
PuroMAG™ Protein L Coated Magnetic Beads, 1 mL
PuroMAG™ Prpotein L coated magnetic beads are designed for efficient capture of IgG and other antibodies from serum, cells, and other biological samples.
Protein L coupled PuroMAG  superparamagnetic beads are suitable for fast and efficient capture of a wide range of Ig’s. Protein L shows varying affinity for Ig’s from different species and for different isotypes within a species (see the Table in the Info section).
During the purification the sample containign Ig's is first mixed with PuroMAG Protein L coated beads. During 30 min incubation the Ig’s bind to PuroMAG-PRL beads via their Fc region. The samples are then placed on a magnetic rack for pull-down and washhing to produce a purified compelex of antibodies bound to Protein L beads. Bound antibodies can then be covalently cross linked to bead-immobilized Protein L to get a very stable bead-antibody complex for fur
Catalog No. 50-256-6975