Learn More
USA Scientific Inc Protein LoBind Tubes, 15 mL
Supplier:  USA Scientific Inc 40112216
Encompass
Protein LoBind Tubes, Protein LoBind, 15 mL, conical tube, PCR clean, colorless, 200 tubes (4 bags × 50 tubes)
Catalog No. 50-274-5320