Learn More
ACROBiosystems Star Ribbon Pre-stained Protein Marker (2.7-40 kDa)
Supplier: ACROBiosystems PPMM1002500UL
This product is a mixture of nine highly purified pre-stained proteins ranging from 2.7 kDa to 40 kDa. The 40 kDa band is orange, 10 kDa band is green, and other bands are blue. It is designed for observing protein separation during SDS-PAGE, verifying western transfer efficiency on membranes, and approximating the size of proteins.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.