Learn More
Sino Biological Assay Buffer and Co-factors: Protein MetalloPhosphatase Buffer (10X)
Supplier:  Sino Biological P240920
Encompass_Preferred
The 10X reaction buffer used for lambda protein phosphatase activity assays.
Catalog No. 50-256-8320