Learn More
Sino Biological Recombinant Mouse Adenylate Kinase 1 Protein (His Tag), Endotoxin-Free 1
Supplier:  Sino Biological 53710M07HUE1
Encompass_Preferred
A DNA sequence encoding the Mouse Adenylate Kinase 1 (NP_032368.2) (Glu35-Gln305) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-006-99036