Learn More
Sino Biological Protein Mouse CHIT1 1MG
Supplier:  Sino Biological 5A2422M08HUE1
Encompass_Preferred
A DNA sequence encoding the Mouse CHIT1 (Q9D7Q1) (Ala22-Ser464) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-98996