Learn More
Sino Biological Protein Mouse Clec9a 100UG
Supplier:  Sino Biological 51145M07HUE100
Encompass_Preferred
A DNA sequence encoding the Mouse Clec9a (NP_001192292.1) (Ser135-Ile264) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-006-98693