Learn More
Sino Biological Recombinant Mouse IL-10 Protein (His Tag), HPLC-verified 100 µg
Supplier:  Sino Biological 50245M07H100
Encompass_Preferred
A DNA sequence encoding the Mouse IL-10 (NP_034678.1) (Ser19-Ser178) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-006-56803