Learn More
Sino Biological Recombinant Mouse Oncostatin M/OSM Protein, Biotinylated 20 µg
Supplier:  Sino Biological 50112MNAEB20
Encompass_Preferred
A DNA sequence encoding the Mouse Oncostatin M/OSM (P53347) (Ala24-Arg206) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-006-56491