Learn More
Sino Biological Protein Mouse PIGR ET-FREE 1MG
Supplier:  Sino Biological 50119M08HUE1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Lys 645) of mouse PIGR (NP 035212 2) precursor was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-73730