Learn More
Sino Biological Recombinant Mouse STING Protein (136-end, His & TEV Tag) 100 µg
Supplier:  Sino Biological S7231H100
Encompass_Preferred
Recombinant mouse STING protein (136-378) was expressed in E. coli cells using an N-terminal His-TEV tag. The protein accession number is Q3TBT3.
Catalog No. 50-006-56562