Learn More
Sino Biological Recombinant Mouse TLR6 Protein (hFc Tag) 100 µg
Supplier:  Sino Biological 50648M02B100
Encompass_Preferred
A DNA sequence encoding the Mouse TLR6 (Q9EPW9) (Phe28-Thr584) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-006-56582