Learn More
Sino Biological Protein Mouse TNFSF15 100UG
Supplier:  Sino Biological 5A7685M07H1UE100
Encompass_Preferred
A DNA sequence encoding the Mouse TNFSF15 (Q5UBV8) (Ile76-Leu252) was expressed, with a polyhistidine tag at the N-terminus. This construct has a trimer design.
Catalog No. 50-006-99031