Learn More
Waters Corp Protein-Pak SEC Glass Column, 10 μm, 8 mm x 300 mm
Supplier: Waters Corp WAT011786
This Protein-Pak SEC Glass Column primarily supports legacy methods. It is constructed with diol-bonded silica-based gel filtration material.
- Chemistry: SEC
- Separation mode: SEC/GPC
- Particle substrate: Silica
- pH range: 2 to 8 pH
- Bonding technology: Diol (OH)
- Silanol activity: Low
- Molecular weight range: 100 to 50000
- Particle shape: Spherical
- Particle size: 10 μm
- Endfitting type: Parker-style
- Pore size: 200 Å
- Format: AP Glass Column
- System: HPLC
- Technique: SEC
- USP classification: L20
- Inner diameter: 8 mm
- Length: 300 mm
- Application: Protein
- Units per package: 1 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.