Learn More
Waters Corp Protein-Pak Hi Res HIC Column, 2.5µm, 4.6 mm x 100 mm, 1/pk
Supplier:  Waters Corp 186007583
Encompass_Preferred
Protein-Pak Hi Res Hydrophobic Interaction Columns (HIC) is ideally suited for use in the characterization of recombinant proteins, monoclonal antibodies, and / or ADCs. The use of non-porous, particles containing a low density of butyl ligands yield outstanding resolution in less time compared to use of many traditional porous HIC offerings. In addition, quality control testing with defined protein standards helps ensure consistent batch-to-batch performance.
Catalog No. 50-152-8581