Learn More
Waters Corp Protein-Pak Column, 10 μm, 8 mm x 300 mm, Glass
Supplier:  Waters Corp WAT011787
Encompass_Preferred
The Protein-Pak 300SW Glass Column is designed to primarily support legacy methods for protein applications. It utilizes 10 μm diol-bonded silica-based gel filtration material.
  • Separates using size exclusion chromatography (SEC) and gel permeation chromatography (GPC)
  • Features silica particle substrate with a spherical shape and low silanol activity
  • Offers a pH range of 2 to 8
  • Utilizes diol (OH) bonding technology
  • Designed for molecular weights ranging from 10000 to 500000
  • Equipped with Parker-style endfittings
  • Pore size of 300 Å
  • Provided in an AP glass column format
  • Packed with water as the solvent
  • Compatible with HPLC systems
  • USP Classification L20
Catalog No. 50-785-050