Learn More
Cayman Chemical Protein Phosphatase 2A C subunit (human recombinant; L309 deletion), 10 µg
SDP
Supplier:  Cayman Chemical 1001123710
Encompass_Preferred
Source: Human recombinant PP2A catalytic subunit expressed in insect cells with an N-terminal octahistidine-tag followed by a streptactin-tag. The C-terminal leucine 309 was deleted. • MW: 38.6 kDa • PP2A is a divalent cation-independent protein serine/threonine phosphatase involved in regulating numerous cellular processes including the cell cycle, growth, and differentiation and is also thought to be a potential tumor suppressor.
Catalog No. NC0314406