Learn More
Cusabio Technology LLC Protein S100-A4 S100A4
Supplier:  Cusabio Technology LLC CSBEL020632MO
Encompass_Preferred
This kit detects S100A4 in serum plasma cell culture supernates and tissue homogenates providing a reliable quantitative analysis within the detection range of 62 5 pg/mL to 4000 pg/mL Utilizing a sandwich ELISA methodology it achieves a sensitivity of 15 6 pg/mL ensuring precise measurement of low-abundance targets The assay requires 50-100 L of sample per test and can be completed in 1-5 hours depending on protocol optimization Results are determined by measuring optical density at 450 nm offering a standardized approach for researchers studying S100A4 expression in diverse biological matrices Its robust design supports applications in both routine laboratory testing and experimental research settings
Catalog No. 50-000-48663