Learn More
BioVendor, LLC Protein Sincalide, 5mg
SDP
Supplier:  BioVendor, LLC RP1782740005
Encompass
Sincalide, a synthetic protein, also called CCK-8 has a molecular formula of C49H62N10O16S3 having an Mw of 1143.29 Dalton. Greater than 99.0% as determined by RP-HPLC.
Catalog No. 50-893-548