Learn More
IBI SCIENTIFIC 3P Proteinase K Powder, 95% Purity, 100 mg
Supplier:  IBI SCIENTIFIC 3P IB05400
Encompass_Preferred
Proteinase K - 100mg IBI Proteinase K is a stable > 50mg/ml (power) highly active proteolytic enzyme that is purified from the mold Tritrachium album.
Catalog No. NC0753471