Learn More
Alkali Scientific Proteinase K, 25 Mg
Supplier: Alkali Scientific OTE001CP
Proteinase K is a highly active protease enzyme commonly used in molecular biology applications, particularly for DNA and RNA extraction. It catalyzes the breakdown of proteins, allowing for the efficient release of nucleic acids from cellular structures during lysis. With its high activity and broad specificity, Proteinase K is effective even in the presence of detergents and other inhibitors commonly used in DNA extraction protocols. Available in a 25 mg package, this reagent is ideal for small-scale experiments or specific laboratory applications where protein degradation is required. Proteinase K is a key enzyme in high-efficiency nucleic acid isolation protocols and enzyme inactivation procedures.