Learn More
Proteintech Group Inc Proteintech Group Antibody, Rabbit Recombinant, Ly-6G, Unconjugated, Mouse Reactivity

Supplier: Proteintech Group Inc 982841RR100UG
Ly-6G (lymphocyte antigen 6 complex, locus G), also known as Gr-1, is a 21-25 kDa, glycosylphosphatidylinositol-anchored protein expressed on myeloid lineage cells in mouse bone marrow. The expression of Ly-6G increases on neutrophils as they differentiate from immature cells in the bone marrow to mature cells in the blood and spleen. This antibody specifically recognizes Ly-6G, but not Ly-6C.
- Recombinant antibody
- Uni-rAb® recombinant antibody
- Specifically recognizes Ly-6G, but not Ly-6C
- Tested for flow cytometry (FC)
- Recommended dilution for FC: 0.25 μg per 106 cells in 100 μl suspension
- Cited for immunohistochemistry (IHC)
- Mouse reactivity
- Unconjugated
- Liquid form
- Purified by protein A
- Stable for one year when stored at 2°C to 8°C
- Storage buffer is PBS with 0.09% sodium azide, pH 7.3
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.