Learn More
Proteintech Group Inc PROTEINTECH Recombinant Monoclonal Antibody, ENO1/2/3, Rabbit IgG, Unconjugated, Liquid

Supplier: Proteintech Group Inc 815041RR
This recombinant monoclonal antibody targets ENO1/2/3 for use in WB, IHC, IF/ICC, FC (Intra), and ELISA, reacting with human, mouse, rat, and pig samples. Enolase (ENO) is a metalloenzyme with three isoforms: ENO1, ENO2, and ENO3. This antibody recognizes all three and binds to an epitope between residues 245 and 258.
- Targets ENO1/2/3.
- Rabbit IgG host/isotype.
- Reacts with human, mouse, rat, pig.
- Validated for WB, IHC, IF/ICC, FC (Intra), ELISA.
- Immunogen: recombinant human ENO1 protein.
- MW: 47 kDa (calculated/observed).
- Unconjugated liquid.
- Protein A purified.
- Storage buffer: PBS, 0.02% sodium azide, 50% glycerol (pH 7.3).
- Stable for one year at -20°C.
- Concentration: 500 μg/mL.
- Size: 100 μL.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.