Learn More
Bioworld Proteomic Grade Detergent Pack, 10 PK(s)
Supplier:  Bioworld 401300051
Encompass_Preferred
bio-World offers a Proteomic Grade Detergent Variety Pack that contains a selection of non-ionic detergents for trial and optimization.The trial pack contains ten 10ml vials of 10% aqueous solution, oneof each of the following proteomic grade detergents:Octyl-D-thioglucopyranoside, CAPS, CHAPS, Tween 20, Tween80, Nonidet P-40, Triton X-100, Triton X-114, Brij 35 and Brij 58. Appearance: Liquid
Catalog No. 50-255-346