Learn More
Sepax Technologies Inc (402NP2-2001) HPLC Column, Sepax, Proteomix WCX-NP1.7, 1.7um, NP 2 x 10 mm
SDP
Supplier:  Sepax Technologies Inc 402NP22001
Encompass
Analytical and Preparative Ion-Exchange Chromatography from Sepax Technologies, Inc. || Proteomix ion-exchange phases have been innovatively developed for achieving high ion-exchange capacity, a breakthrough technology for non-porous resins. Proteomix packing support is composed of a rigid, spherical, highly cross-linked poly(styrene divinylbenzene) (PS/DVB) non-porous bead with particle size selections of 1.7, 3, 5 and 10 µm. The hydrophobic PS/DVB resin surface is totally covered by a hydrophilic coating that eliminates non-specific bindings with biological analytes, leading to high efficiency and high recovery separations for biological molecules. On the top of the hydrophilic layer, ion-exchange functional groups are attached via a proprietary chemistry, resulting in a high capacity ion-exchange layer. For full product details, additional dimensions, rush orders, and custom requests, please contact the manufacturer.
Catalog No. 50-320-5340