Learn More
ATCC Proteus mirabilis
Supplier:  ATCC 7002
Encompass
Proteus mirabilis was isolated from the urine of a human patient with kidney stones. The bacterial culture has applications in quality control, urinary tract infection research, media testing, and food testing.
Catalog No. 50-238-1958