Learn More
Medchemexpress LLC Protocatechuate 3,4-dioxygenase | 9029-47-4 | ≥3 U/mg solid | 100 U
SDP
Supplier:  Medchemexpress LLC HYP2915100U
Encompass_Preferred
Protocatechuate 3,4-dioxygenase is a dioxygenase belonging to the non-heme iron dioxygenase class. It catalyzes the cleavage of the aromatic ring of 3,4-dihydroxybenzoate by attaching two atoms of molecular oxygen to generate β-carboxy-cis,cis-muconate. This enzyme is a key component in the β-ketoadipic acid pathway and can be found in marine bacteria associated with Roseobacter, and can be isolated from Pseudomonas aeruginosa.
  • Dioxygenase that catalyzes aromatic ring cleavage
  • Key enzyme in the β-ketoadipic acid pathway
  • Found in marine bacteria associated with Roseobacter
  • Can be isolated from Pseudomonas aeruginosa
  • Used as a biochemical assay reagent
  • Involved in anti-infection pathways
Catalog No. 50-003-98703