Learn More
Medchemexpress LLC Protocatechuate 3,4-dioxygenase | 9029-47-4 | 50 U
SDP
Supplier:  Medchemexpress LLC HYP291550U
Encompass_Preferred
Protocatechuate 3,4-dioxygenase is a dioxygenase that belongs to the non-heme iron dioxygenase class. This enzyme catalyzes the cleavage of the aromatic ring of 3,4-dihydroxybenzoate by incorporating two atoms of molecular oxygen to generate β-carboxy-cis,cis-muconate. It is a key enzyme in the β-ketoadipic acid pathway, found in marine bacteria associated with Roseobacter, and can be isolated from Pseudomonas aeruginosa.
  • Catalyzes the aromatic ring cleavage of 3,4-dihydroxybenzoate.
  • Generates β-carboxy-cis,cis-muconate.
  • Key enzyme in the β-ketoadipic acid pathway.
  • Found in marine bacteria associated with the Roseobacter group.
  • Can be isolated from Pseudomonas aeruginosa.
Catalog No. 50-003-98706