Learn More
Medchemexpress LLC Protodioscin | 55056-80-9 | 1049.20 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN079910MM/1ML
Encompass_Preferred
Protodioscin is a major steroidal saponin found in *Trigonella foenum-graecum Linn.* It exhibits multiple biological actions, including anti-hyperlipidemia, anti-cancer properties, sexual effects, and cardiovascular benefits. It is for research use only.
  • Multiple biological actions
  • Anti-hyperlipidemia properties
  • Anti-cancer properties
  • Sexual effects
  • Cardiovascular benefits
  • For research use only
Catalog No. 50-004-16933