Learn More
Medchemexpress LLC Protodioscin | 55056-80-9 | 1049.20 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN079950MG
Encompass_Preferred
Protodioscin is a major steroidal saponin found in Trigonella foenum-graecum Linn. It exhibits a wide range of biological activities, including anti-hyperlipidemia, anti-cancer, sexual effects, and cardiovascular properties. This compound has been shown to improve glucose intolerance and reduce various serum and renal markers of inflammation and disease progression.
  • Exhibits anti-hyperlipidemia, anti-cancer, sexual, and cardiovascular effects.
  • Cytotoxic against human 293T and HUVEC cell lines (IC50 = 3.22 μM).
  • Improves glucose intolerance and reduces serum levels of UA, BUN, Cr, TC, and TG.
  • Reduces renal concentrations of IL-1β, IL-6, and TNF-α.
  • Ameliorates death rate and neurological deficits in MCAO rats.
  • Increases coagulation time by approximately 50%.
  • Lowers LDL and increases HDL/LDL ratio.
Catalog No. 50-004-16230