Learn More
Medchemexpress LLC Protoescigenin | 20853-07-0 | 5 MG

Supplier: Medchemexpress LLC HYN74975MG
Protoescigenin is the main aglycone of horse chestnut saponin mixture known as escin. Protoescigenin is selected as substrate for exploratory chemistry towards selective protection, followed by propargyl ether formation and subsequent condensation with azido-monosaccharides, to obtain novel triazole linked conjugates of the triterpene.
- Purity: 98.82%
- Molecular weight: 506.71
- Formula: C30H50O6
- Appearance: Solid
- Color: White to off-white
- Structure classification: Terpenoids, Triterpenes
- Initial source: Plants, other families
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.