Learn More
Apexbio Technology LLC Protopanaxdiol 30636-90-9 20mg
SDP
Supplier:  Apexbio Technology LLC N159720
Encompass_Preferred
Protopanaxdiol (CAS 30636-90-9) is a dammarane-type triterpenoid saponin aglycone derived from Panax ginseng C A Mey This compound exerts multiple biological effects through modulation of cellular signaling pathways including anti-inflammatory and anticancer activities via regulation of apoptosis and inhibition of tumor cell proliferation Protopanaxdiol is frequently utilized in preclinical research to investigate the pharmacological mechanisms of ginsenosides and to explore its potential therapeutic roles in oncology neurology and immunology The molecule is soluble in acetone and DMSO and should be stored in a tightly sealed container under cool dry conditions
Catalog No. 50-000-90702