Learn More
Medchemexpress LLC Protopine | 130-86-9 | 353.37 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN079310MM/1ML
Encompass_Preferred
Protopine (Corydinine) is an isoquinoline alkaloid that acts as a specific reversible and competitive inhibitor of acetylcholinesterase. It also demonstrates anti-inflammatory, anti-microbial, anti-angiogenic, and anti-tumour activities.
  • Specific reversible and competitive inhibitor of acetylcholinesterase
  • Exhibits anti-inflammation, anti-microbial, anti-angiogenic, and anti-tumour activity
Catalog No. 50-004-17088