Learn More
Medchemexpress LLC Protopine | 130-86-9 | 353.37 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0793100MG
Encompass_Preferred
Protopine (Corydinine) is an isoquinoline alkaloid and a specific, reversible, and competitive inhibitor of acetylcholinesterase. This compound exhibits diverse biological activities, including anti-inflammatory, anti-microbial, anti-angiogenic, and anti-tumour effects. It has been shown to inhibit cell viability, migration, and invasion in liver carcinoma cells and induce apoptosis.
  • Inhibits acetylcholinesterase
  • Possesses anti-inflammatory, anti-microbial, and anti-angiogenic properties
  • Demonstrates anti-tumour activity by affecting cell viability, migration, and apoptosis
  • Modulates the PI3K/Akt signaling pathway
  • Alleviates memory impairment in animal models
  • Exhibits antidepressant-like effects in vivo
  • Provides protective effects against cerebral ischaemic injury
Catalog No. 50-004-16814