Learn More
Medchemexpress LLC Protopine | 130-86-9 | 353.37 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN079350MG
Encompass_Preferred
Protopine (Corydinine) is an isoquinoline alkaloid that acts as a specific reversible and competitive inhibitor of acetylcholinesterase. It exhibits anti-inflammatory, anti-microbial, anti-angiogenic, and anti-tumor activities.
  • Inhibits cell viability, migration, invasion, and EMT process in liver carcinoma cells.
  • Induces apoptosis by increasing caspase-3 and caspase-9 expression.
  • Inhibits the PI3K/Akt signaling pathway.
  • Induces ROS generation.
  • Inhibits the uptake of serotonin transporter (SERT) and noradrenaline (NE).
  • Alleviates scopolamine-induced memory impairment.
  • Inhibits tumor growth in xenograft mice.
  • Shows antidepressant-like effects.
  • Exhibits protective effects on focal cerebral ischemic injury.
Catalog No. 50-004-16103