Learn More
Creative Biomart Hemin
Supplier:  Creative Biomart THP0073
Encompass_Preferred
The product is an iron-containing porphyrin. More specifically, it is protoporphyrin IX containing a ferric iron ion (heme B) with a chloride ligand.
Catalog No. 50-208-4124