Learn More
Medchemexpress LLC Protoporphyrin IX (disodium) | 50865-01-5 | 96.5% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1247A
Encompass_Preferred
Protoporphyrin IX disodium is a final intermediate in the heme biosynthetic pathway. It acts as a radiation sensitizer, enhancing ROS generation even in hypoxic conditions and inducing DNA damage. It also functions as a photosensitizer, undergoing photobleaching through direct degradation by light irradiation. This product is promising for research of sonodynamic and photodynamic agents for various cancers, including bladder cancer and nodular basal cell carcinoma.
  • Final intermediate in the heme biosynthetic pathway
  • Acts as a radiation sensitizer
  • Enhances ROS generation and induces DNA damage
  • Functions as a photosensitizer
  • Undergoes photobleaching by light irradiation
  • Accumulates in tumor cells after 5-aminolevulinic acid administration
  • Causes selective improvement of basal cell carcinoma
  • Promising for research into sonodynamic and photodynamic agents for various cancers
Catalog No. 50-004-13842