Learn More
Medchemexpress LLC Protoporphyrin IX disodium | 50865-01-5 | 96.5% | 606.62 | C34H32N4Na2O4 | 10 MG

Supplier: Medchemexpress LLC HYB1247A10MG
Protoporphyrin IX disodium is the disodium salt of protoporphyrin IX, supplied for laboratory research including studies of heme biosynthesis and photodynamic or sonodynamic therapy. It is provided as a solid with analytical documentation to support research use.
- CAS number 50865-01-5
- Molecular formula C34H32N4Na2O4
- Molecular weight 606.62
- Purity 96.5%
- Appearance solid, brown to black
- Available in small research quantities, including 10 MG
- Documentation available: SDS and COA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.