Learn More
Medchemexpress LLC Protoporphyrin IX (Standard) | 553-12-8 | 96.1% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB1247R50MG
Encompass_Preferred
Protoporphyrin IX (Standard) serves as an analytical reference standard for diverse research and analytical applications. It functions as a key intermediate in the heme biosynthetic pathway, acting as both a radiation sensitizer-boosting reactive oxygen species (ROS) generation and inducing DNA damage even in hypoxic states-and a photosensitizer, undergoing photobleaching upon light exposure.
  • Enhances ROS generation and induces DNA damage.
  • Undergoes photobleaching through direct degradation by light irradiation.
  • Accumulates in tumor cells after 5-aminolevulinic acid (5-ALA) administration.
  • Causes selective improvement of basal cell carcinoma when activated by red fluorescence.
  • Promising for research into sonodynamic and photodynamic agents for various cancers.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Ideal for applications in HPLC, GC, and MS.
Catalog No. 50-004-12812