Learn More
Carbosynth LLC. ProTX-II, 5mg, For a Smaller Quantity in Acetate Form, See PTX-4450-s, Disulfide bonds: Disulfide bonds between Cys2-Cys16, Cys9-Cys21, and Cys15-Cys25, MW: 3826.66, Molecular formula: C168H250N46O41S8, CAS# 484598-36-9
SDP
Supplier:  Carbosynth LLC. PTX3995PI5MG
Encompass_Preferred
Synonyms: H-Tyr-Cys-Gln-Lys-Trp-Met-Trp-Thr-Cys-Asp-Ser-Glu-Arg-Lys-Cys-Cys-Glu-Gly-Met-Val-Cys-Arg-Leu-Trp-Cys-Lys-Lys-Lys-Leu-Trp-OH, H-YCQKWMWTCDSERKCCEGMVCRLWCKKKLW-OH, Application: Na+ Channel (Especially Nav1.7) / Ca2+ Channel Blocker (Gating Modifier), Salt Form:, Trifluoroacetate, Storage Temperature: -20 °C, References: R.E. Middleton et al., Biochemistry, 41,14734 (2002)., J.J. Smith et al., J. Biol. Chem., 282, 12687 (2007)., W. A. Schmalhofer et al., Mol. Pharmacol., 74, 1476 (2008).
Catalog No. 50-168-6803