Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 54 (d15B) 2mg
Supplier:  ATCC 518X
Encompass
Purified pneumococcal polysaccharide type 15B is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 15B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-0959