Learn More
Aviva Systems Biology Corp Prox2 Antibody - N-terminal region (ARP37543_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37543_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prox2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the n terminal region of mouse Prox2. Alternate names: 1700058C01RikApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-654